Learn More
Epredia™ Cover Slips, Rectangles
Epredia™TM coverslips are ideally suited as cover material for specimens in microscopic examinations in medicine, biology and research. Manufactured from D 263TM M borosilicate glass from Schott AG. Available in various sizes and thicknesses.
66.80 € - 319.00 €
Product Code | Brand | Dimensions (L x W) | Thickness (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Thickness (Metric) | Price | Quantity & Availability | ||||
11936309
|
Epredia™
BB02400360AC13MNZ0 |
36 x 24 mm | #1.5 (0.16 to 0.19 mm) |
66.80 €
Pack of 1000 |
Estimated Shipment: 03-07-2024
Log in to see stock. |
||||
12765098
|
Epredia™
A67761330 |
64 x 48 mm | #1 (0.13 to 0.16 mm) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Highly transparent and achromatic – practically free of any color distortion
- Good planarity
- Refraction index adjusted for microscopes
- High optical homogeneity – practically free of inclusions, streaks, bubbles etc.
- Very good chemical stability
Specifications
Rectangle | |
100/Pk. |