missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CRTC3 Recombinant Rabbit Monoclonal Antibody (K01_1L06)

Product Code. 30830413
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830413 50 μL
30825422 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830413 Supplier Invitrogen™ Supplier No. MA564752

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

CRTC3 Recombinant Monoclonal Antibody for ICC/IF, Western Blot

The protein encoded by this gene has similarity to a yeast protein. It seems to be part of a peripheral membrane protein complex localized on cis/medial Golgi cisternae where it may participate in tethering intra-Golgi transport vesicles.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRTC3
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone K01_1L06
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 0.05% BSA, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene Crtc3
Gene Accession No. Q6UUV7, Q91X84
Gene Alias 2610312F20Rik; 6332415K15Rik; AI429792; BC011210; CREB regulated transcription coactivator 3; CREB-regulated transcription coactivator 3; Crtc3; RGD1309666; TORC3; TORC-3; transducer of CREB protein 3; transducer of regulated cAMP response element-binding protein (CREB) 3; transducer of regulated cAMP response element-binding protein 3; transducer of regulated CREB protein 3
Gene Symbols Crtc3
Host Species Rabbit
Immunogen A synthetic peptide of human CRTC3.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 365297, 64784, 70461
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.