Learn More
Corning™ Cryogenic Vial Grippers
Enables to easily sort or move vials maintaining sterility. Corning™ Cryogenic Vial Grippers feature unique design to grasp internal or external-thread cryogenic vials.
Brand:  Corning™ 432136
Code : NEW
Additional Details : Weight : 0.10350kg
Product Code. 15565535
  • 54.10 € / Pack of 5
Estimated Shipment: 11-09-2024
to see stock.
Add to basket
Description
Description
Cryogenic vial grippers feature a unique design to grasp internal or external-thread cryogenic vials. Grippers enable to easily sort or move vials while maintaining sterility and protecting fingers from frozen vials, dry ice and liquid nitrogen. 5 per case.
Specifications
Specifications
5/Pk.
Vial Grippers
Sorting and moving cryogenic vials
15.2 cm