Learn More
Technical Service Consultants™ Cryopreservation kit
Protect™ provides a cost effective means of storing microorganisms
Brand:  Technical Service Consultants™ TS73-DY 25
Code : 03
Additional Details : Weight : 1.00000kg
Product Code. 12897755
  • 34.40 € / Pack of 25
Call for latest local availability
01 885 5854
Description
Description
Protect™ provides a cost effective means of storing microorganisms. Each vial contains 20 beads in a cryopreserve solution, to keep the DNA identity of the organism intact.
  • Ceramic beads for improved recovery and minimal cell loss
  • Flat cap top offers easy identification
Anaerobe
  • Developed for problematic organisms
  • Increased viscosity and oxygen removal
  • Essential trace elements
  • Improved viability and recovery
    Dairy
  • Ideal media to maintain milk and dairy microorganisms
  • Based on BS and ISO recommendations
  • Suitable for water and environmental isolates
    Meat-free
  • Pharmaceutical and bio-fermentation
  • TSE-free alternative
  • Full product security
  • Reduced vessel completion time
    Yeast and mould
  • Developed with a spore 'grab' formulation
  • Easy suspension of spores and yeast cells
  • Neutral pH for increased cell stability
Cryopreservation of bacterial, yeast and mould samples.
Specifications
Specifications
Cryopreservation Kit
Ceramic beads, plasticized paper box ideal media to maintain milk and diary organisms TSC