Learn More
Saint Gobain Life Sciences Tygon™ LMT-55 Tubing with Three Stop Configuration
Formulated for resistance to flex fatigue and abrasion and is less permeable than rubber tubing. Tygon™ LMT-55 Tubing with Three Stop Configuration handles virtually all inorganic chemicals found in the lab.
Brand:  Saint Gobain Life Sciences 070535-11-ND
Code : NEW
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 13474815
  • 70.45 € / Pack of 12
In Stock
to see stock.
Add to basket
Description
Description
  • Bio-based formulation delivers excellent performance
  • Lot-to-lot consistency for reproducible results
  • Non-toxic and non-contaminating
  • Smooth, polished inner wall
  • Slips easily over fittings and grips securely for simple lab set-ups
  • Contains no BPA or phthalates
  • Standard sizes available to hold full vacuum at room temperature
Typical Applications
  • General laboratory
  • Food and beverage
  • Biopharmaceutical
  • Analytical instruments
  • Peristaltic and vacuum pumps
  • Ideal for condensers, incubators, desiccators, gas lines and drain line
Specifications
Specifications
Tubing
White/White
1.02 mm
0.11 in.
32.8 ft.
0.03 in.
For sample and reagent dispensing, biological and chemical assays, and water and soil analysis
1 Ea.
Crystal Clear
Flexible
FDA Compliant, NSF, 3-A, USP Class VI
0.04 in.
Tygon R3607
2.72 mm
10 m
0.85 mm
-50°C
1.02 x 2.72 mm
55 (Shore A)
74°C