Learn More
Cole-Parmer™ CrystalVu™ Luer Fitting with Male Lueur Lock x 1/8 in. Hose Barb
Available in 6 colors for quick and easy identification: berry, rose, yellow, lime, violet and orange
Brand: Cole-Parmer™ 45520-23
Code : NEW
Additional Details : Weight : 0.04000kg
Description
- Made of polyolefin, a transparent polycarbonate alternative containing no bisphenol-A (BPA).
- Superior chemical resistance compared to polycarbonate.
- Sterilize by autoclave, gamma irradiation or ethylene oxide.
- Meets USP Class VI.
- Animal-derived ingredient-free.
- Available in packs of 25.
Specifications
Luer fitting, male luer lock x 1/8 in. hose barb | |
Polyolefin | |
Yellow |
Male luer lock | |
25/Pk. |