missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CSNK1E (CK1 epsilon) R178C Recombinant Human Protein
This CSNK1E (CK1 epsilon) R178C Recombinant Human Protein (full length) was expressed in insect cells.
Brand: Invitrogen™ A33498
Additional Details : Weight : 0.10000kg
Description
- This CSNK1E (CK1 epsilon) R178C recombinant human protein (full length) was expressed in insect cells
- CSNK1E serine/threonine kinase is a member of the CK1 family which plays an important role in diverse cell processes, including DNA replication and repair
- It is a regulator of Yes-associated protein (YAP) transcription coactivator, which is a key regulator of organ size and a candidate human oncogene
- CSNK1E is activated by CCK2R and then phosphorylates PKD2 at Ser244
- Phosphorylation of PKD2 leads to its nuclear accumulation, and efficient phosphorylation of nuclear PKD2 substrates in human gastric cancer cells
- CSNK1E can phosphorylate topoisomerase (topo) II alpha at serine-1106 which regulates the enzyme activity and sensitivity to topo II-targeted drugs
- CSNK1E (CK1 epsilon) R178C recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service
- Validated for use with the Z'-LYTE™ assay
- Available in multiple pack sizes for convenient use
Specifications
NP_001885.1 | |
10 µg | |
Full Length | |
GST Tag | |
Dry Ice | |
Kinases & Inhibitors | |
Liquid | |
Human |
73.6 kDa | |
CK1 Kinase Family | |
Yes | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol | |
Baculovirus | |
Full Length, Recombinant, Mutant | |
Kinase Assay |