missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CSPD™ Substrate (0.25 mM Ready-To-Use)

Product Code. 10655886
Click to view available options
Quantity:
50 mL, 50mL
100 mL, 100mL
250 mL
Unit Size:
50mL
Each
This item is not returnable. View return policy

Product Code. 10655886

Brand: Invitrogen™ T2141

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This CSPD™ substrate is a chemiluminescent alkaline phosphatase substrate, ready-to-use for protein or nucleic acid blotting on nitrocellulose membranes.

This CSPD™ substrate is a chemiluminescent alkaline phosphatase substrate, ready-to-use for protein or nucleic acid blotting on nitrocellulose membranes. This substrate can also be used in solution-based assays.

This CSPD™ chemiluminescent substrate for alkaline phosphatase let you detect alkaline phosphatase and alkaline phosphatase-labeled molecules with unparalleled sensitivity, speed, and ease.

  • Chemiluminescent substrates provide highly sensitive replacements for the widely used fluorogenic substrate methylumbelliferyl phosphate (MUP), and the colorimetric substrate p-nitrophenyl phosphate (pNPP).
  • Low background luminescence coupled with high-intensity light output enables detection of alkaline phosphatase labels with the highest possible sensitivity and signal-to-noise.

This versatile chemiluminescent substrate exhibits high sensitivity in membrane-based applications such as Southern, Northern, and Western blotting. This substrate can also be used in solution-based assays such as immunoassays, DNA probe assays, enzyme assays, and reporter gene assays. Maximum light levels are reached in approximately 10 minutes and glow emission persists for several hours.

TRUSTED_SUSTAINABILITY

Specifications

Product Type ELISA Substrate, CSPD™ Substrate
Quantity 50 mL, 50mL
Detection Method Chemiluminescence
Form Liquid
For Use With (Application) Protein assays and analysis
Format Bottle
Substrate Type AP (Alkaline Phosphatase) Substrate, Alkaline Phosphatase
Product Line CSPD™, NovaBright™
Item Description CSPD™ Substrate, 50mL, 0.25mM Concentration, Liquid, AP (Alkaline Phosphatase) Substrate Type, Room Temperature Shipping Condition, 4°C Storage Requirements, Used For Protein Assays and Analysis
Packaging 1 Each
Content And Storage Store the product at 4°C.
Shipping Condition Room Temperature
Storage Requirements 4°C
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.