missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CSPD™ Substrate (0.4 mM Ready-To-Use) with Sapphire-II™ Enhancer, Size B
Click to view available options
Quantity:
100 ml
Unit Size:
Each
Description
Chemiluminescent substrates provide highly sensitive replacements for the widely used fluorogenic substrate methylumbelliferyl phosphate (MUP), and the colorimetric substrate p-nitrophenyl phosphate (pNPP). Low background luminescence coupled with high-intensity light output enables detection of alkaline phosphatase labels with the highest possible sensitivity and signal-to-noise.
- Detect alkaline phosphatase and alkaline phosphatase-labeled molecules with unparalleled sensitivity, speed and ease
- Exhibits high sensitivity in membrane-based applications such as Southern, Northern, and Western blotting
- Use in solution-based assays such as immunoassays, DNA probe assays, enzyme assays, and reporter gene assays
- Maximum light levels are reached in approximately 10 minutes and glow emission persists for several hours
- Enhancers increase light output and sensitivity
Specifications
Specifications
| Product Type | ELISA Substrate |
| Quantity | 100 ml |
| Detection Method | Chemiluminescence |
| Form | Liquid |
| Format | Bottle |
| Product Line | CSPD™, NovaBright™ |
| Substrate Type | Alkaline Phosphatase |
| Content And Storage | Store the product at 4°C. |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction