missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cullin 2 Recombinant Rabbit Monoclonal Antibody (K01_1P92)

Product Code. 30830411
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830411 50 μL
30826988 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830411 Supplier Invitrogen™ Supplier No. MA564884

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Cullin 2 Recombinant Monoclonal Antibody for Western Blot

Core component of multiple cullin-RING-based ECS (ElonginB/C-CUL2/5-SOCS-box protein) E3 ubiquitin-protein ligase complexes, which mediate the ubiquitination of target proteins. ECS complexes and ARIH1 collaborate in tandem to mediate ubiquitination of target proteins (PubMed:27565346). May serve as a rigid scaffold in the complex and may contribute to catalysis through positioning of the substrate and the ubiquitin-conjugating enzyme. The E3 ubiquitin-protein ligase activity of the complex is dependent on the neddylation of the cullin subunit and is inhibited by the association of the deneddylated cullin subunit with TIP120A/CAND1. The functional specificity of the ECS complex depends on the substrate recognition component. ECS(VHL) mediates the ubiquitination of hypoxia-inducible factor (HIF). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cullin 2
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1P92
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 0.05% BSA, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene Cul2
Gene Accession No. Q13617, Q9D4H8
Gene Alias 1300003D18Rik; 4932411N15Rik; AI327301; CUL2; CUL-2; cullin 2; cullin-2; MGC131970; mKIAA4106; OTTHUMP00000019437; testis secretory sperm-binding protein Li 238E
Gene Symbols Cul2
Host Species Rabbit
Immunogen A synthetic peptide of human Cullin 2.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361258, 71745, 8453
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.