Learn More
Hellma™ Suprasil™ Quartz TrayCell
Designed for the UV/Vis analysis of DNA/RNA and proteins. Hellma™ Suprasil™ Quartz TrayCells have dimensions equivalent to a standard cuvette in order to work in most spectrophotometers.
Brand: Hellma™ 105810-A3-V1-46
Code : 50
Additional Details : Weight : 0.30000kg
Description
- Extremely flexible and cost-effective solution for the analysis of very small sample volumes
- Ideal for biomolecular laboratories, to perform the analysis of nucleic acids and proteins in very small volumes
- Determination of labeling efficiency for microarray experiments
- All UV/Vis analysis utilizing the wavelength range of 190 to 1100nm
- Quick and easy cleaning
- No dilution of sample needed
- No evaporation of the sample through the cap
Specifications
0.7 to 5μL |