Learn More
Hellma™ Cuvette ultra micro cell far uv quartz light path 10mm, centre height 8.5mm blue 20μL
Brand:  Hellma™ 105-204-1085-40
Code : 33
Additional Details : Weight : 1.00000kg
Product Code. 12771695