missing translation for 'onlineSavingsMsg'
Learn More

Cyanoacetic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-4229152
Click to view available options
Quantity:
25 g
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy
372-09-8
C3H3NO2
85.06
MLIREBYILWEBDM-UHFFFAOYSA-N
cyanoacetic acid, acetic acid, cyano, malonic mononitrile, acide cyanacetique, cyanessigsaeure, monocyanoacetic acid, malonic acid mononitrile, usaf kf-17, cyanoethanoic acid, kyselina kyanoctova
9740
CHEBI:51889
2-cyanoacetic acid
C(C#N)C(=O)O
This item is not returnable. View return policy
372-09-8
C3H3NO2
85.06
MLIREBYILWEBDM-UHFFFAOYSA-N
cyanoacetic acid, acetic acid, cyano, malonic mononitrile, acide cyanacetique, cyanessigsaeure, monocyanoacetic acid, malonic acid mononitrile, usaf kf-17, cyanoethanoic acid, kyselina kyanoctova
9740
CHEBI:51889
2-cyanoacetic acid
C(C#N)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 65.0°C to 67.0°C
Color White to Beige
Boiling Point 108.0°C (15.0 mmHg)
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula NCCH2CO2H
Quantity 25 g
Beilstein 02,583
Fieser 11,145; 13,171
Merck Index 15,2679
Solubility Information Solubility in water: 1000g/L (20°C). Other solubilities: soluble in alcohol and ether,slightly soluble in benzene and chloroform
Formula Weight 85.06
Percent Purity 97%
Physical Form Adhering Crystalline Solid
Chemical Name or Material Cyanoacetic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'