missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyanogen iodide, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 142.0°C to 149.0°C |
Color | Beige to Brown or Pink |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (Iodometry) |
Packaging | Glass bottle |
Linear Formula | ICN |
Quantity | 5 g |
Merck Index | 15,2685 |
Solubility Information | Solubility in water: soluble in water. Other solubilities: soluble in diethyl ether,ethanol,ether,alcohols,and volatile oils |
Molecular Weight (g/mol) | 152.92 |
Show More |
Safety and Handling
Product Identifier
- Cyanogen iodide
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H301-Toxic if swallowed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'