Learn More
Contec™ CyChlor
Provided ready-to-use and is efficacious against bacteria and yeasts in 3 mins. Contec™ CyChlor is a new broad spectrum disinfectant designed for every day use.
Brand: Contec™ FBC100CC
75.63 EUR valid until 2024-04-30
Use promo code "21673" to get your promotional price.
Additional Details : Weight : 6.50000kg
Description
- Broad spectrum efficacy in 3 mins saves time spent on biodecontamination.
- Filtered to 0.2 microns and filled in a Grade A environment ensuring the product is free from contamination and particulates
- Contains no quaternary ammonium or surfactant so it's very low residue, saving time on residue removal
- No hazard classification so only basic PPE required and no special disposal is necessary
- Trigger spray can be set to jet or spray - large droplet size reduces the risk of inhalation and provides good surface coverage
- Double bagged packed in linear tear packaging so each bag is easy to open even when wearing gloves
- Not classed as corrosive and can be used safely in all areas of the cleanroom – this needs a bit of random text deleting from the end
Specifications
Non-sterile | |
No | |
1 L |
Capped Container | |
6/Cs. |