missing translation for 'onlineSavingsMsg'
Learn More

Cyclobutanemethanol, 99%, Thermo Scientific Chemicals

Catalog Number p-96388
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
4415-82-1
C5H10O
86.13
MFCD00001330
WPOPOPFNZYPKAV-UHFFFAOYSA-N
cyclobutanemethanol, hydroxymethylcyclobutane, cyclobutylcarbinol, hydroxymethyl cyclobutane, cyclobutyl methanol, cyclobutyl-methanol, cyclobutanecarbinol, cyclobutane carbinol, pubchem23465
78130
cyclobutylmethanol
C1CC(C1)CO
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
4415-82-1
C5H10O
86.13
MFCD00001330
WPOPOPFNZYPKAV-UHFFFAOYSA-N
cyclobutanemethanol, hydroxymethylcyclobutane, cyclobutylcarbinol, hydroxymethyl cyclobutane, cyclobutyl methanol, cyclobutyl-methanol, cyclobutanecarbinol, cyclobutane carbinol, pubchem23465
78130
cyclobutylmethanol
C1CC(C1)CO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Density 0.9100g/mL
Boiling Point 143°C to 144°C
Flash Point 40°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C4H7CH2OH
Refractive Index 1.4450 to 1.447
Quantity 5 g
Specific Gravity 0.91
Solubility Information Solubility in water: immiscible
Formula Weight 86.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Cyclobutanemethanol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Cyclobutanemethanol
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'