Learn More
Cyclobutanemethanol, 99%, ACROS Organics™
Brand: Acros Organics 182230050
Code : 29
Additional Details : CAS Number : 4415-82-1 Weight : 0.05500kg Transport : UN number : 1987 Chem class : 3 Pack group : III
Packaging | Glass bottle |
---|---|
Quantity | 5g |
Chemical Identifiers
4415-82-1 | |
86.13 | |
WPOPOPFNZYPKAV-UHFFFAOYSA-N | |
78130 | |
C1CC(C1)CO |
C5H10O | |
MFCD00001330 | |
cyclobutanemethanol, hydroxymethylcyclobutane, cyclobutylcarbinol, hydroxymethyl cyclobutane, cyclobutyl methanol, cyclobutyl-methanol, cyclobutanecarbinol, cyclobutane carbinol, pubchem23465 | |
cyclobutylmethanol |
Specifications
Cyclobutanemethanol | |
40°C | |
Glass bottle | |
0.91 | |
Colorless | |
99% | |
99% | |
C4H7CH2OH | |
cyclobutanemethanol, hydroxymethylcyclobutane, cyclobutylcarbinol, hydroxymethyl cyclobutane, cyclobutyl methanol, cyclobutyl-methanol, cyclobutanecarbinol, cyclobutane carbinol, pubchem23465 | |
WPOPOPFNZYPKAV-UHFFFAOYSA-N | |
cyclobutylmethanol | |
78130 | |
Liquid |
0.9100g/mL | |
Authentic | |
1.4450 to 1.447 | |
143°C to 144°C | |
5g | |
4415-82-1 | |
C5H10O | |
MFCD00001330 | |
Solubility in water: immiscible | |
C1CC(C1)CO | |
86.13 | |
86.13 | |
99% |
Safety and Handling
- Cyclobutanemethanol
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.