Learn More
cis-Cyclooctene, 95%, stabilized, ACROS Organics™
Brand: Acros Organics 154862500
Code : 29
Additional Details : CAS Number : 931-87-3 Weight : 0.26000kg Transport : UN number : 3295 Chem class : 3 Pack group : III
Packaging | Glass bottle |
---|---|
Quantity | 250mL |
Chemical Identifiers
931-87-3 | |
110.2 | |
cis-cyclooctene, z-cyclooctene, cyclooctene, z, cyclooctene, 1z, cyclooct-1-ene, e-cyclooctene, 1-cyclooctene #, 1z-cyclooctene, 7z-cyclooctene, ar,e-cyclooctene | |
cyclooctene |
C8H14 | |
URYYVOIYTNXXBN-UPHRSURJSA-N | |
638079 | |
C1CCCC=CCC1 |
Specifications
cis-Cyclooctene | |
25°C | |
Glass bottle | |
0.84 | |
1.89 mPa.s (20°C) | |
Colorless to Brown | |
250mL | |
94% min. (GC) | |
C8H14 | |
Solubility in water: <0.1g/L (20°C) | |
C1CCCC=CCC1 | |
110.2 | |
110.2 | |
95% |
0.8400g/mL | |
Authentic | |
1.4680 to 1.4710 | |
100 to 200ppm irganox 1076 FD | |
145.0°C to 146.0°C | |
-16.0°C | |
931-87-3 | |
C8H14 | |
cis-cyclooctene, z-cyclooctene, cyclooctene, z, cyclooctene, 1z, cyclooct-1-ene, e-cyclooctene, 1-cyclooctene #, 1z-cyclooctene, 7z-cyclooctene, ar,e-cyclooctene | |
URYYVOIYTNXXBN-UPHRSURJSA-N | |
cyclooctene | |
638079 | |
Liquid |