missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanecarbonyl chloride, 98%, ACROS Organics™

Brand:  Acros Organics 354371000

Code : 29

Additional Details : CAS Number : 4524-93-0 Weight : 0.10000kg Transport : UN number : 2920 Chem class : 8 Pack group : II


Packaging Glass bottle
Quantity 100g

 View more versions of this product

Product Code. 11934971

  • 511.00 € / 100g

Estimated Shipment: 08-07-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

4524-93-0
132.59
cyclopentanecarbonylchloride, cyclopentane carboxyl chloride, cyclopentanoyl chloride, pubchem13237, acmc-1csoo, cyclopentylformyl chloride, cyclopetanecarbonylchloride, cyclopentancarbonyl chloride, cyclopentylcarbonyl chloride, chlorocarbonyl cyclopentane
cyclopentanecarbonyl chloride
C6H9ClO
WEPUZBYKXNKSDH-UHFFFAOYSA-N
78284
C1CCC(C1)C(=O)Cl

Specifications

Cyclopentanecarbonyl chloride
60°C
Glass bottle
1.091
Colorless to Yellow
4524-93-0
C6H9ClO
WEPUZBYKXNKSDH-UHFFFAOYSA-N
cyclopentanecarbonyl chloride
78284
Liquid
1.0910g/mL
Authentic
1.4610 to 1.4640
54.0°C to 56.0°C (15.0mmHg)
100g
98%
cyclopentanecarbonylchloride, cyclopentane carboxyl chloride, cyclopentanoyl chloride, pubchem13237, acmc-1csoo, cyclopentylformyl chloride, cyclopetanecarbonylchloride, cyclopentancarbonyl chloride, cyclopentylcarbonyl chloride, chlorocarbonyl cyclopentane
C1CCC(C1)C(=O)Cl
132.59
132.59
98%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Cyclopentanecarbonyl chloride

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS Documents
Certificates