Learn More
Cyclopentanecarboxaldehyde, 97%, stabilized, ACROS Organics™
Brand: Acros Organics 425910250
Code : 29
Additional Details : CAS Number : 872-53-7 Weight : 0.07500kg Transport : UN number : 1989 Chem class : 3 Pack group : III
Packaging | Glass bottle |
---|---|
Quantity | 25g |
Chemical Identifiers
872-53-7 | |
98.14 | |
cyclopentanecarboxaldehyde, cyclopentanealdehyde, cyclopentancarbaldehyd, cyclopentylformaldehyde, formylcyclopentane, cyclopentyl aldehyde, 1-formylcyclopentane, cyclopentane-1-carboxaldehyde, cyclopentanecaboxaldehyde, cyclopentanecarboaldehyde | |
cyclopentanecarbaldehyde |
C6H10O | |
VELDYOPRLMJFIK-UHFFFAOYSA-N | |
70106 | |
C1CCC(C1)C=O |
Specifications
Cyclopentanecarboxaldehyde | |
28°C | |
Glass bottle | |
0.919 | |
140.0°C to 141.0°C | |
25g | |
96% min. (GC) | |
C5H9CHO | |
VELDYOPRLMJFIK-UHFFFAOYSA-N | |
cyclopentanecarbaldehyde | |
70106 | |
Liquid |
0.9190g/mL | |
Authentic | |
1.4420 to 1.4460 (20°C, 589nm) | |
0.1 wt% hydroquinone | |
Colorless to Yellow | |
872-53-7 | |
C6H10O | |
cyclopentanecarboxaldehyde, cyclopentanealdehyde, cyclopentancarbaldehyd, cyclopentylformaldehyde, formylcyclopentane, cyclopentyl aldehyde, 1-formylcyclopentane, cyclopentane-1-carboxaldehyde, cyclopentanecaboxaldehyde, cyclopentanecarboaldehyde | |
C1CCC(C1)C=O | |
98.14 | |
98.14 | |
97% |
Safety and Handling
- Cyclopentanecarboxaldehyde
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.