Learn More
Cyclosporin A, 99+%, Thermo Scientific™
Cyclosporin A, CAS # 59865-13-3, is a compound that shows immunosuppressive, antifungal, antirheumatic, and dermatologic activities. It is also a calcineurin and enzyme inhibitor.
217.00 € - 486.00 €
Chemical Identifiers
| CAS | 59865-13-3 |
|---|---|
| Molecular Formula | C62H111N11O12 |
| Molecular Weight (g/mol) | 1202.64 |
| MDL Number | MFCD00274558 |
| InChI Key | PMATZTZNYRCHOR-IMVLJIQENA-N |
| Synonym | Cyclosporine A; Antibiotic S 7481F1 |
| PubChem CID | 132274082 |
| IUPAC Name | 30-ethyl-33-[(4E)-1-hydroxy-2-methylhex-4-en-1-yl]-1,4,7,10,12,15,19,25,28-nonamethyl-6,9,18,24-tetrakis(2-methylpropyl)-3,21-bis(propan-2-yl)-1,4,7,10,13,16,19,22,25,28,31-undecaazacyclotritriacontan-2,5,8,11,14,17,20,23,26,29,32-undecone |
| SMILES | CCC1NC(=O)C(C(O)C(C)C\C=C\C)N(C)C(=O)C(C(C)C)N(C)C(=O)C(CC(C)C)N(C)C(=O)C(CC(C)C)N(C)C(=O)C(C)NC(=O)C(C)NC(=O)C(CC(C)C)N(C)C(=O)C(NC(=O)C(CC(C)C)N(C)C(=O)CN(C)C1=O)C(C)C |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15475449
|
Thermo Scientific Alfa Aesar
J63191.03 |
1g |
217.00 €
1g |
In Stock
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
15485449
|
Thermo Scientific Alfa Aesar
J63191.06 |
5g |
486.00 €
5g |
In Stock
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 59865-13-3 | |
| 1202.64 | |
| PMATZTZNYRCHOR-IMVLJIQENA-N | |
| 132274082 | |
| CCC1NC(=O)C(C(O)C(C)C\C=C\C)N(C)C(=O)C(C(C)C)N(C)C(=O)C(CC(C)C)N(C)C(=O)C(CC(C)C)N(C)C(=O)C(C)NC(=O)C(C)NC(=O)C(CC(C)C)N(C)C(=O)C(NC(=O)C(CC(C)C)N(C)C(=O)CN(C)C1=O)C(C)C |
| C62H111N11O12 | |
| MFCD00274558 | |
| Cyclosporine A; Antibiotic S 7481F1 | |
| 30-ethyl-33-[(4E)-1-hydroxy-2-methylhex-4-en-1-yl]-1,4,7,10,12,15,19,25,28-nonamethyl-6,9,18,24-tetrakis(2-methylpropyl)-3,21-bis(propan-2-yl)-1,4,7,10,13,16,19,22,25,28,31-undecaazacyclotritriacontan-2,5,8,11,14,17,20,23,26,29,32-undecone |
Specifications
| 59865-13-3 | |
| C62H111N11O12 | |
| 3647785 | |
| Cyclosporine A; Antibiotic S 7481F1 | |
| PMATZTZNYRCHOR-IMVLJIQENA-N | |
| 30-ethyl-33-[(4E)-1-hydroxy-2-methylhex-4-en-1-yl]-1,4,7,10,12,15,19,25,28-nonamethyl-6,9,18,24-tetrakis(2-methylpropyl)-3,21-bis(propan-2-yl)-1,4,7,10,13,16,19,22,25,28,31-undecaazacyclotritriacontan-2,5,8,11,14,17,20,23,26,29,32-undecone | |
| 132274082 | |
| ≥99% |
| White | |
| MFCD00274558 | |
| 14,2752 | |
| Soluble in dimethyl sulfoxide and ethanol; Insoluble in water. | |
| CCC1NC(=O)C(C(O)C(C)C\C=C\C)N(C)C(=O)C(C(C)C)N(C)C(=O)C(CC(C)C)N(C)C(=O)C(CC(C)C)N(C)C(=O)C(C)NC(=O)C(C)NC(=O)C(CC(C)C)N(C)C(=O)C(NC(=O)C(CC(C)C)N(C)C(=O)CN(C)C1=O)C(C)C | |
| 1202.64 | |
| 1202.61 | |
| Cyclosporin A |
Safety and Handling
GHS H Statement
H361-H302
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
H302-H350-H360FD
missing translation for 'einecsNumber' : 611-907-1
missing translation for 'rtecsNumber' : GZ4120000
missing translation for 'tsca' : No
Recommended Storage : Store at -20°C
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.