Learn More
Lenz Laborglasinstrumente™ Cylindrical Dropping Funnels
For the safe, easy and accurate transfer of fluids in the laboratory. Lenz Laborglasinstrumente™ Cylindrical Dropping Funnels are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ B0496701
Additional Details : Weight : 0.14100kg
Product Code. 10050540
  • 135.00 € / Each
Estimated Shipment: 01-07-2024
to see stock.
Add to basket
Description
Description
  • With inscription label
  • With NS socket and cone
  • With scale (approx. volume/mL)
  • With pressure equalizing tube
  • With screwthread retaining device
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
50 mL
Duran Glass, Borosilicate Glass
Pressure Equalization Tube at the Side