missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CYM 50260
GREENER_CHOICE

Product Code. 30001536
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001536

Brand: Tocris Bioscience™ 4677/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective S1P4 agonist

Potent sphingosine-1-phosphate receptor 4 (S1P4) agonist (EC50 = 45 nM). Displays no activity at other S1P receptor subtypes (S1P1-3 and S1P5) at concentrations up to 25 μM.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1355026-60-6
Molecular Formula C14H11Cl3FNO2

Specifications

Quantity 50 mg
Formula Weight 350.6
Purity >98%
Molecular Weight (g/mol) 350.595
Chemical Name or Material CYM 50260
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.