Learn More
Invitrogen™ Cystatin 2 Monoclonal Antibody (237305)
Rat Monoclonal Antibody
Brand:  Invitrogen™ MA560995
Product Code. 30535019
  • 589.00 € / 500µg
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
The cystatin superfamily is a well-established family of cysteine protease inhibitors. Cystatins A and B (type 1) are mainly intracellular; cystatins C, D, E/M, F, G, S, SN and SA cystatins are extracellular (type 2); and the kininogens are type 3 cystatins which are intravascular proteins. All true cystatins inhibit cysteine peptidases of the papain family, such as cathepsins, and some also inhibit legumain family enzymes. Cystatin SA, cystatin S and cystatin SN are found primarily in saliva. Cystatin S and SN can also be expressed in tears, urine and seminal fluid. Cystatin C is a related protein which is expressed in brain, thymus, ovary, epididymis and vas deferens. Cystatin D protects against proteinases in the oral cavity, while Cystatin E/M and F moderate the inhibition of cathepsin proteins. The fetuins, part of the cystatin superfamily, are secretable proteins that influence osteogenesis and bone resorption, regulation of the insulin and hepatocyte growth factor receptors and the response to systemic inflammation. High molecular weight kininogen (Kininogen HC) and low molecular weight kininogen (Kininogen LC) have varied roles, though they both inhibit the thrombin- and plasmin-induced aggregation of thrombocytes.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cystatin 2
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Rat
Protein A/G
RUO
Mouse
Antibody
IgG2b
Immunoprecipitation, Western Blot
237305
Unconjugated
CST2; cystatin 2; cystatin S5; cystatin SA; cystatin-2; Cystatin-S5; cystatin-SA; cysteine-proteinase inhibitor; salivary cysteine (thiol) protease inhibitor
E. coli-derived recombinant mouse Cystatin/Stefin Homolog 2, Met2-Phe97.
500 μg
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized