missing translation for 'onlineSavingsMsg'
Learn More

BioTek Instruments Cytation C10 Confocal Imaging Reader

The Cytation C10 Confocal Imaging Reader combines automated digital confocal and widefield microscopy with conventional multi-mode microplate reading in a unique, patented design. The spinning disk confocal module provides exquisite resolution and optical sectioning capabilities in a wide variety of sample types. 

Brand:  BioTek Instruments C10PW

Additional Details : Weight : 53.30000kg

 View more versions of this product

Product Code. 17160569

  • 78877.00 € / 1 unit

Estimated Shipment: 21-09-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • High quality components used in the system, including a Hamamatsu scientific CMOS (sCMOS) camera, Olympus objectives and laser-based illumination enable excellent image quality, at an affordable price
  • Includes widefield fluorescence, brightfield and phase contrast optics
  • Its variable bandwidth monochromator-based multi-mode plate reading is based on the proven design of the market-leading BioTek Synergy™ products
  • Environmental controls enable live cell assays, and Gen5™ software is specifically designed to make sample detection and image capture quick and effortless
  • The new 3D viewer provides users with 3D reconstruction of their thicker samples captured with the confocal microscope.
  • The Cytation C10 brings affordable confocal to every laboratory
  • Confocal imaging and multi-mode plate reader in one
  • Environmental controls for live cell imaging
  • Hit-picking: Imaging and multi-mode detection saves time and data storage
  • Variable bandwidth for sensitivity and specificity
  • Cytation C10 is ready for any assay
Note:  250 lb. shipping weight. White glove inside delivery with material lift required for all orders unless exempted; contact your Fisher Scientific Sales Representative for more details.
Specifications

Specifications

230 to 999 nm
Monochromator 4 nm (230 to 285 nm), 8 nm (>285 nm)
20 x 27 x 18.5 (D x W x H)
7 decades fluorescence >6 decades luminescence 0 to 4 OD
50/60 Hz
2 syringe optional
Monochromator: 6- to 384-well plates. Imaging 6 to 1536 well plates
UV Vis absorbance, Flourescence intensity, Luminescence
Linear, orbital, double orbital
Gen5 Microplate Reader and Imager Software included. Gen5 Secure for 21 CFR Part 11 compliance (option). Gen5 Image+ and Image Prime software available for full image analysis (option)
122 lb.
27 in.
OD accuracy <1% (at 3 OD)
Range: 0 to 20% (CO2); 1 to 19% (O2), with optional Gas Controller
5 to 1000 µL in 1 µL increments (optional with injectors)
Instrument external 250 W, 24 VDC power supply, compatible with 100-240 VAC (at 50-60 Hz)
C10PW Configuration
1 to 19% with optional gas controller
12 Months
4-Zone incubation to 45°C with Condensation Control
Xenon Flash
100/240 V
53.3 kg
68.6 cm
Videos
SDS
Documents
Certificates
View accessory products