missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 6A (KRT6A) (Basal Cell Marker) Mouse, Clone: KRT6A/2368, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio 3853-MSM4-P1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15990318

  • 459.10 € / 100µg

Estimated Shipment: 03-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Cytokeratin 6A (KRT6A) (Basal Cell Marker)
Cytoplasmic
200μg/mL
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide.
Recombinant full-length human Cytokeratin 6A (KRT6A) protein
56kDa
Protein A/G purified from Bioreactor concentrate.
RUO
Primary
3853
Immunohistochemistry
KRT6A/2368
Unconjugated
Mouse
IgG2a, kappa
Tonsil or Basal Cell Carcinoma (BCC)
100 μg
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Monoclonal
Videos
SDS
Documents
Certificates