missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin, Acidic Mouse, Clone: KRTL/1377, enQuireBio™
Mouse Monoclonal Antibody
199.00 € - 485.00 €
Specifications
Antigen | Cytokeratin, Acidic |
---|---|
Clone | KRTL/1377 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15991398
|
EnquireBio
MSM24-1377-P1 |
100 μg |
485.00 €
100µg |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15981398
|
EnquireBio
MSM24-1377-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Cytokeratin, Acidic | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
374454 | |
Recombinant human KRT77 protein fragment | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
the usefulness of keratins as markers in cancer research and tumor diagnosis. |
KRTL/1377 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
XP_011536590 | |
KRT77 | |
IgG1 | |
Protein A/G Purified | |
Cytoplasmic | |
Skin, Squamous cell carcinoma (SCC) |