missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin, Acidic Mouse, Clone: KRTL/1377, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio MSM24-1377-P1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15991398

  • 459.10 € / 100µg

Estimated Shipment: 03-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Cytokeratin, Acidic
Cytoplasmic
200μg/mL
10mM PBS with 0.05% BSA and 0.05% Azide
KRT77
Recombinant human KRT77 protein fragment
the usefulness of keratins as markers in cancer research and tumor diagnosis.
Protein A/G Purified
RUO
Primary
374454
Immunohistochemistry
KRTL/1377
Unconjugated
XP_011536590
Mouse
IgG1
Skin, Squamous cell carcinoma (SCC)
100 μg
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Monoclonal
Videos
SDS
Documents
Certificates