missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Cytometric Bead Array (CBA) Functional Beads
Color-coded 7.5μm polystyrene beads that can be classified using a flow cytometer
1270.00 € - 1470.00 €
Specifications
Content And Storage | 4°C |
---|---|
Contents | Aqueous buffered solution containing ≤0.09% sodium azide |
For Use With (Application) | Flow Cytometry |
Material | Polystyrene |
Product Type | Functional Bead |
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
15839498
|
BD
558578 |
Functional Bead |
1270.00 €
Each |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15859498
|
BD
558580 |
Functional Bead |
1340.00 €
Each |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15879498
|
BD
558582 |
Functional Bead |
1440.00 €
Each |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15869498
|
BD
558581 |
Functional Bead |
1355.00 €
Each |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15889498
|
BD
558583 |
Functional Bead |
1440.00 €
Each |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15849498
|
BD
558579 |
Functional Bead |
1355.00 €
Each |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15899498
|
BD
558584 |
Functional Bead |
1425.00 €
Each |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15809508
|
BD
558585 |
Functional Bead |
1465.00 €
Each |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15819508
|
BD
558586 |
Functional Bead |
1420.00 €
Each |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15849548
|
BD
558645 |
Functional Bead |
1425.00 €
Each |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15869548
|
BD
558647 |
Functional Bead |
1335.00 €
Each |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15889548
|
BD
558649 |
Functional Bead |
1470.00 €
Each |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15899548
|
BD
558650 |
Functional Bead |
1410.00 €
Each |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15829558
|
BD
558656 |
Functional Bead |
1335.00 €
Each |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15889868
|
BD
560031 |
Functional Bead |
1335.00 €
Each |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15899868
|
BD
560032 |
Functional Bead |
1440.00 €
Each |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15829878
|
BD
560035 |
Functional Bead |
1405.00 €
Each |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15819878
|
BD
560034 |
Functional Bead |
1425.00 €
Each |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15809878
|
BD
560033 |
Functional Bead |
1405.00 €
Each |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15839878
|
BD
560036 |
Functional Bead |
1425.00 €
Each |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15849878
|
BD
560037 |
Functional Bead |
1405.00 €
Each |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15869878
|
BD
560039 |
Functional Bead |
1405.00 €
Each |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15879878
|
BD
560040 |
Functional Bead |
1455.00 €
Each |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15879548
|
BD
558648 |
Functional Bead |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15839558
|
BD
558657 |
Functional Bead |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15829508
|
BD
558587 |
Functional Bead |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15859548
|
BD
558646 |
Functional Bead |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
The Functional Beads can be used in conjunction with the BD CBA Functional Bead Conjugation Buffer Set (Mfr. No. 558556) to covalently link water-soluble proteins to the surface of the beads via a sulfo-SMCC chemistry. The beads, once linked to a protein, can be used as a capture matrix for analytes of interest.
Specifications
4°C | |
Flow Cytometry | |
Functional Bead | |
6000 | |
7.5 μm |
Aqueous buffered solution containing ≤0.09% sodium azide | |
Polystyrene | |
Flow cytometer | |
0.9 mL |