missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Cytometric Bead Array Flex Set Standard
Used as standard control
160.00 € - 169.00 €
Specifications
Content And Storage | 4°C |
---|---|
Product Type | Cytometric Bead Array (CBA) Standard |
Format | Lyophilized |
Includes | Lyophilized recombinant Human Interferon-l (IFN-l) |
Quantity | 0.2 mL |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15889398
|
BD
558456 |
Human IFN-gamma Flex Set Standard |
166.00 €
Each |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15899398
|
BD
558457 |
Human IL-1beta Flex Set Standard |
160.00 €
Each |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15869408
|
BD
558464 |
Human IL-6 Flex Set Standard |
169.00 €
Each |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15849408
|
BD
558462 |
Human IL-4 Flex Set Standard |
166.00 €
Each |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15839408
|
BD
558461 |
Human IL-2 Flex Set Standard |
166.00 €
Each |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15809408
|
BD
558458 |
Human IL-10 Flex Set Standard |
166.00 €
Each |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15849418
|
BD
558472 |
Human TNF Flex Set Standard |
166.00 €
Each |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15879408
|
BD
558465 |
Human IL-8 Flex Set Standard |
166.00 €
Each |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15869418
|
BD
558474 |
Mouse IL-10 Flex Set Standard |
164.00 €
Each |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15819428
|
BD
558479 |
Mouse IL-6 Flex Set Standard |
160.00 €
Each |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15829428
|
BD
558480 |
Mouse TNF Flex Set Standard |
160.00 €
Each |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This product contains lyophilized recombinant human Interferon-λ (IFN-λ)
- Storage Buffer: Lyophilized in aqueous buffered solution containing BSA and ProClin™ 150
Flow cytometry
Specifications
4°C | |
Lyophilized | |
0.2 mL |
Cytometric Bead Array (CBA) Standard | |
Lyophilized recombinant Human Interferon-l (IFN-l) | |
RUO |