missing translation for 'onlineSavingsMsg'
Learn More

Cytosine β-D-arabinofuranoside hydrochloride, 99%, crystalline, ACROS Organics™

Brand:  Acros Organics 227180050

Code : 29

Additional Details : CAS Number : 69-74-9 Weight : 0.00500kg


Packaging Glass bottle
Quantity 5g

 View more versions of this product

Product Code. 10021704

  • 570.60 € / 5g

Estimated Shipment
08-10-2020

Please to purchase this item. Need a web account? Register with us today!



Specifications

Chemical Identifiers

69-74-9
279.67
KCURWTAZOZXKSJ-JCEIVDPDSA-N
134129637
C1=CN(C(=O)N=C1N)C2C(C(C(O2)CO)O)O.Cl
C9H13N3O5·HCl
MFCD00012839
cytarabine hydrochloride
4-amino-1-[(2S,3R,4S,5S)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one;hydrochloride

Specifications

Cytosine β-D-arabinofuranoside hydrochloride
Glass bottle
White
5g
98.5% min. (HPLC)
MFCD00012839
cytarabine hydrochloride
C1=CN(C(=O)N=C1N)C2C(C(C(O2)CO)O)O.Cl
279.67
279.67
99%
Authentic
+127° to +133° (20°C, 589nm) ((c=0.7, H2O))
197.0°C to 198.0°C
69-74-9
C9H13N3O5·HCl
10, 2778
KCURWTAZOZXKSJ-JCEIVDPDSA-N
4-amino-1-[(2S,3R,4S,5S)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one;hydrochloride
134129637
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Cytosine beta-D-arabinofuranoside hydrochloride

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
  • Respiratory sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H361d-Suspected of damaging the unborn child.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents