missing translation for 'onlineSavingsMsg'
Learn More

Cytosine-β-D-arabinofuranoside Hydrochloride, Fisher BioReagents

Catalog Number p-25274
Click to view available options
Quantity:
100 mg
500 mg
5 g
Packaging:
Amber Glass
This item is not returnable. View return policy
69-74-9
C9H14ClN3O5
279.68
MFCD00012839
KCURWTAZOZXKSJ-JBMRGDGGSA-N
cytarabine hydrochloride
134129637
4-amino-1-[(2S,3R,4S,5S)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one;hydrochloride
Cl.NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](CO)[C@@H](O)[C@@H]1O
This item is not returnable. View return policy
69-74-9
C9H14ClN3O5
279.68
MFCD00012839
KCURWTAZOZXKSJ-JBMRGDGGSA-N
cytarabine hydrochloride
134129637
4-amino-1-[(2S,3R,4S,5S)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one;hydrochloride
Cl.NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](CO)[C@@H](O)[C@@H]1O
Cytosine-β-D-Arabinofuranoside Hydrochloride inhibits incorporation of labeled thymidine into DNA.
TRUSTED_SUSTAINABILITY
Melting Point 197°C
Color White
Packaging Amber Glass
Quantity 100 mg
Formula Weight 279.7
Physical Form Powder or Solid
Chemical Name or Material Cytosine-β-D-Arabinofuranoside Hydrochloride
compliance-icons compliance-icons
Product Identifier
  • Cytosine beta-D-arabinofuranoside hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H361d-Suspected of damaging the unborn child.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'