missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals D-Alanine, 99%
27.40 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Description
D-Alanine is essential for the biosynthesis of peptidoglycan crosslinking sub-units that are used for bacterial cell walls.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
D-Alanine is essential for the biosynthesis of peptidoglycan crosslinking sub-units that are used for bacterial cell walls.
Solubility
Soluble in water (155 mg/ml at 20°C), ethanol (slightly), and pyridine (slightly). Insoluble in ether, and acetone.
Notes
Store in cool, dry conditions in well sealed containers. Strong oxidizing agents.
D-Alanine is essential for the biosynthesis of peptidoglycan crosslinking sub-units that are used for bacterial cell walls.
Solubility
Soluble in water (155 mg/ml at 20°C), ethanol (slightly), and pyridine (slightly). Insoluble in ether, and acetone.
Notes
Store in cool, dry conditions in well sealed containers. Strong oxidizing agents.
Specifications
Melting Point | ∼295°C (decomposition) |
Beilstein | 1720249 |
Merck Index | 14,204 |
Solubility Information | Soluble in water (155mg/ml at 20°C),ethanol (slightly),and pyridine (slightly). Insoluble in ether,and acetone. |
Formula Weight | 89.09 |
Percent Purity | 99% |
Quantity | 5 g |
Optical Rotation | −14° (c=5 in 5N HCl) |
Chemical Name or Material | D-Alanine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'