missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(+)-Cellobiose, 98%

Catalog Number p-3735816
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
528-50-7
C12H22O11
342.30
MFCD00136034
GUBGYTABKSRVRQ-UHFFFAOYNA-N
d-+-cellobiose, 4-o-beta-glucopyranosyl-d-glucose
57370084
(3S,4S,6S)-2-(hydroxymethyl)-6-[(3S,4R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O
This item is not returnable. View return policy
528-50-7
C12H22O11
342.30
MFCD00136034
GUBGYTABKSRVRQ-UHFFFAOYNA-N
d-+-cellobiose, 4-o-beta-glucopyranosyl-d-glucose
57370084
(3S,4S,6S)-2-(hydroxymethyl)-6-[(3S,4R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 239°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 31, 380
Merck Index 15, 1963
Specific Rotation + 34
Solubility Information (5% in water) Clear colorless
Formula Weight 342.29
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material D(+)-Cellobiose
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'