missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-(+)-Glucose monohydrate

Catalog Number p-7081213
Click to view available options
Quantity:
500 g
2500 g
14431-43-7
C6H12O6·H2O
MFCD00149450
14431-43-7
C6H12O6·H2O
MFCD00149450

D-(+)-Glucose monohydrate is used by industry professionals to indicate the degree to which a substance contains water, and in food industry, pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(+)-Glucose monohydrate is used by industry professionals to indicate the degree to which a substance contains water, and in food industry, pharmaceuticals.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Melting Point ∽83°C
Assay Percent Range 99%
Quantity 500 g
Solubility Information Soluble in water.
Formula Weight 198.18 (180.16anhydrous)
Chemical Name or Material D-(+)-Glucose monohydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'