missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Data Output Accessories for Environmental Chambers and Incubators
Integrate Thermo Scientific™ Data Output Accessories for your environmental chamber or incubator to increase functionality and compatibility.
77.00 € - 3320.00 €
Product Code | Brand | Description | For Use With (Equipment) | Installation | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | For Use With (Equipment) | Installation | Price | Quantity & Availability | |||
15533761
|
Thermo Scientific™
197030 |
Chart paper for single pen chart recorders | Environmental Chambers and Incubators | Customer Installed |
77.00 €
Each |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||
15583761
|
Thermo Scientific™
201147 |
Dual Pen, 6 in. 7 Day Chart Recorders; 0° to 60°C (32° to 140°F) | Environmental Chamber (3948 and 3907) | Customer Installed |
3110.00 €
Each |
Estimated Shipment: 23-08-2024
Log in to see stock. |
|||
15553761
|
Thermo Scientific™
201144 |
Single pen, 6 in. 7 Day Chart Recorders; 0° to 60°C (32° to 140°F) | Environmental Chamber (3948 and 3907) | Factory Installed |
2255.00 €
Each |
Estimated Shipment: 01-08-2024
Log in to see stock. |
|||
15573761
|
Thermo Scientific™
201146 |
Dual Pen, 6 in. 7 Day Chart Recorders; 0° to 60°C (32° to 140°F) | Environmental Chamber (3948 and 3907) | Factory Installed |
3320.00 €
Each |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||
15563761
|
Thermo Scientific™
201145 |
Single Pen, 6 in. 7 Day Chart Recorders; 0° to 60°C (32° to 140°F) | Environmental Chamber (3948 and 3907) | Customer Installed |
2170.00 €
Each |
Estimated Shipment: 23-04-2024
Log in to see stock. |
|||
Description
Integrate Thermo Scientific™ Data Output Accessories for your environmental chamber or incubator to increase functionality and compatibility.
Specifications
LED | |
Accessory, Chart Recorders and Paper |