missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Decafluorobenzophenone, 97%, Thermo Scientific Chemicals
32.86 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
Description
Perfluorobenzophenone was used in the synthesis of fluorinated acridones and acridines. Fluorinated polyetherketones prepared from decafluorobenzophenone. Laser pulse excitation of decafluorobenzophenone (DFB) in acetonitrile yields the triplet 3DFB.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Perfluorobenzophenone was used in the synthesis of fluorinated acridones and acridines. Fluorinated polyetherketones prepared from decafluorobenzophenone. Laser pulse excitation of decafluorobenzophenone (DFB) in acetonitrile yields the triplet 3DFB.
Solubility
Soluble in methanol. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Perfluorobenzophenone was used in the synthesis of fluorinated acridones and acridines. Fluorinated polyetherketones prepared from decafluorobenzophenone. Laser pulse excitation of decafluorobenzophenone (DFB) in acetonitrile yields the triplet 3DFB.
Solubility
Soluble in methanol. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Melting Point | 92°C to 94°C |
Quantity | 1 g |
Beilstein | 2066476 |
Solubility Information | Soluble in methanol. Insoluble in water. |
Formula Weight | 362.12 |
Percent Purity | 98% |
Chemical Name or Material | Decafluorobenzophenone |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'