missing translation for 'onlineSavingsMsg'
Learn More

Decane, 99%, anhydrous, AcroSeal™, Thermo Scientific Chemicals

Catalog Number p-4229161
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
124-18-5
C10H22
142.28
DIOQZVSQGTUSAI-UHFFFAOYSA-N
n-decane, nonane, methyl, decyl hydride, unii-nk85062oiy, ccris 653, hsdb 63, dsstox_cid_4913, decane, analytical standard, dsstox_rid_77577, dsstox_gsid_24913
15600
CHEBI:41808
decane
CCCCCCCCCC
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
124-18-5
C10H22
142.28
DIOQZVSQGTUSAI-UHFFFAOYSA-N
n-decane, nonane, methyl, decyl hydride, unii-nk85062oiy, ccris 653, hsdb 63, dsstox_cid_4913, decane, analytical standard, dsstox_rid_77577, dsstox_gsid_24913
15600
CHEBI:41808
decane
CCCCCCCCCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -30.0°C
Color Colorless
Density 0.7350g/mL
Boiling Point 174.0°C
Flash Point 46°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging AcroSeal™ Glass Bottle
Linear Formula CH3(CH2)8CH3
Refractive Index 1.4103 to 1.4123
Quantity 100 mL
Residue after Evaporation 0.0005 % max.
Specific Gravity 0.735
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol (miscible with ethanol),soluble in ether,slightly soluble in ccl4
Formula Weight 142.28
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Decane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Decane
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Flammable liquid Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H226-Flammable liquid and vapour.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'