missing translation for 'onlineSavingsMsg'
Learn More

Decanophenone, 98+%

Product Code. p-7044520
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11377046

Brand: Thermo Scientific Alfa Aesar L06923.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6048-82-4 | C16H24O | 232.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6048-82-4
Molecular Formula C16H24O
Molecular Weight (g/mol) 232.37
MDL Number MFCD00009582
InChI Key QQXJNLYVPPBERR-UHFFFAOYSA-N
Synonym decanophenone, 1-decanone, 1-phenyl, n-decanophenone, capriphenone, ketone, nonyl phenyl, ketone, nonenyl phenyl, nonyl phenyl ketone, 1-phenyl-1-decanone, acmc-209mkb
PubChem CID 80148
IUPAC Name 1-phenyldecan-1-one
SMILES CCCCCCCCCC(=O)C1=CC=CC=C1

Specifications

Melting Point 34°C to 36°C
Boiling Point 165°C to 167°C (7 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 2048790
Formula Weight 232.37
Percent Purity ≥98%
Chemical Name or Material Decanophenone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.