missing translation for 'onlineSavingsMsg'
Learn More

(+)-Dehydroabietylamine hydrochloride, Thermo Scientific Chemicals

Catalog Number p-7076604
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
16496-99-4
C20H32ClN
321.93
MFCD06795849
CVPQLGCAWUAYPF-UHFFFAOYNA-N
leelamine hydrochloride, dehydroabiethylamine hydrochloride, +-dehydroabietylamine, lylamine hydrochloride, nsc 2955 hydrochloride, +-dehydroabietylamine hydrochloride, +-dehydroabiethylamine hydrochloride, abieta-8,11,13-trien-18-amine-hydrogen chloride 1/1, 13-isopropyl-podocarpa-8,11,13-trien-15-amine hydrochloride, 1r,4as,10ar-7-isopropyl-1,4a-dimethyl-1,2,3,4,4a,9,10,10a-octahydrophenanthren-1-yl methanamine hydrochloride
16759156
[(1R,4aS,10aR)-1,4a-dimethyl-7-propan-2-yl-2,3,4,9,10,10a-hexahydrophenanthren-1-yl]methanamine;hydrochloride
Cl.CC(C)C1=CC2=C(C=C1)C1(C)CCCC(C)(CN)C1CC2
This item is not returnable. View return policy
16496-99-4
C20H32ClN
321.93
MFCD06795849
CVPQLGCAWUAYPF-UHFFFAOYNA-N
leelamine hydrochloride, dehydroabiethylamine hydrochloride, +-dehydroabietylamine, lylamine hydrochloride, nsc 2955 hydrochloride, +-dehydroabietylamine hydrochloride, +-dehydroabiethylamine hydrochloride, abieta-8,11,13-trien-18-amine-hydrogen chloride 1/1, 13-isopropyl-podocarpa-8,11,13-trien-15-amine hydrochloride, 1r,4as,10ar-7-isopropyl-1,4a-dimethyl-1,2,3,4,4a,9,10,10a-octahydrophenanthren-1-yl methanamine hydrochloride
16759156
[(1R,4aS,10aR)-1,4a-dimethyl-7-propan-2-yl-2,3,4,9,10,10a-hexahydrophenanthren-1-yl]methanamine;hydrochloride
Cl.CC(C)C1=CC2=C(C=C1)C1(C)CCCC(C)(CN)C1CC2

Resolving agent for carboxylic acids

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Packaging Plastic bottle
Quantity 10 mg
Beilstein 3084620
Formula Weight 321.9
Physical Form Solid
Chemical Name or Material (+)-Dehydroabietylamine hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'