missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dengue Virus Envelope Antibody (1) [Biotin] - Pan Specific, (Domain III), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-43767B
This item is not returnable.
View return policy
Description
Dengue Virus Envelope Monoclonal antibody specifically detects Dengue Virus Envelope in Virus samples. It is validated for ELISA,Western Blot
Specifications
| Dengue Virus Envelope | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
| ELISA, Western Blot | |
| 1 | |
| Western Blot, ELISA | |
| dengue virus polyprotein | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Dengue Virus Envelope (type 1, US/Hawaii/1944) (RefSeq#: ACF49259.1; Val580-Lys680). | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction