Learn More
Fisherbrand™ Descaling Agent, Citric Acid Base
Fisherbrand™ Descaling Agent, Citric Acid Base is a clear and colorless liquid suitable for use in water baths.
Brand: Fisherbrand™
Additional Details : Weight : 10.00000kg
Description
- Citric acid monohydrate, 2-Hydroxypropane-1,2,3-tri carbonic acid
- Clear, colorless liquid
- pH value at 25°C: 2.2 (10g/L), 1.85 (50g/L)
Specifications
Water Bath | |
Citric acid-based descaling agent | |
5 L |
Descaling | |
At 25°C: 2.2 (10g/L), 1.85 (50g/L) |
Safety and Handling
- Decalcifier / Descaler - Citric Acid based 5kg Canister
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P102-Keep out of reach of children.
- P264-Wash hands and face thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : citric acid monohydrate