missing translation for 'onlineSavingsMsg'
Learn More

Dess-Martin periodinane, 15 wt.% solution in dichloromethane, AcroSeal™, Thermo Scientific Chemicals

Catalog Number 10060498
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

oxidation reagent
TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
CAS 87413-09-0, 75-09-2
Molecular Formula C13H13IO8
MDL Number MFCD00130127
Density 1.3620g/mL
Merck Index 15, 2934
Synonym dess-martin periodinane, triacetoxyperiodinane, 1,1,1-triacetoxy-1,1-dihydro-1,2-benziodoxol-3 1h-one, dess-martin reagent, dess-martinperiodinane, dess-martin, dess martin periodinane, dess martin, dess-martin periodane
InChI Key NKLCNNUWBJBICK-UHFFFAOYSA-N
SMILES CC(=O)O[I]1(OC(C)=O)(OC(C)=O)OC(=O)C2=CC=CC=C12
IUPAC Name (1,1-diacetyloxy-3-oxo-1$l^{5},2-benziodoxol-1-yl) acetate
Molecular Weight (g/mol) 424.14
PubChem CID 159087
Formula Weight 424.15
Percent Purity 10 to 15%
Packaging AcroSeal™ Glass Bottle
Specific Gravity 1.362
Quantity 100 mL
Physical Form Liquid
Chemical Name or Material Dess-Martin periodinane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'