missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-n-butyltin oxide, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
1 kg
Description
Di-n-butyltin oxide is used as a reagent and a catalyst. It is particularly useful in regioselective alkylation, acylation and sulfonation reactions for starting materials containing alcohol functional groups. It is used in studies pertaining to discovering novel antitumor agents, antifungal activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | >300°C |
Color | White |
Density | 1.58 g/mL |
Odor | Odorless |
Linear Formula | [CH3(CH2)3]2SnO |
Quantity | 1 kg |
UN Number | UN3146 |
Beilstein | 4126243 |
Solubility Information | Soluble in water 4.0g/L @ 20°C. |
Formula Weight | 248.92 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'