missing translation for 'onlineSavingsMsg'
Learn More

Di-n-octyltin oxide, Thermo Scientific Chemicals

Catalog Number p-7080334
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
870-08-6
C16H34OSn
361.16
MFCD00013839
LQRUPWUPINJLMU-UHFFFAOYSA-N
di-n-octyltin oxide, stannane, dioctyloxo, dioctyltin oxide, dioctyloxostannane, tin, dioctyloxo, dioctyl oxo stannane, stannane, oxodioctyl, di-n-octyl-zinn oxyd, tin, dioctyl-, oxide, dioctyl oxo tin
93563
dioctyl(oxo)tin
CCCCCCCC[Sn](=O)CCCCCCCC
This item is not returnable. View return policy
870-08-6
C16H34OSn
361.16
MFCD00013839
LQRUPWUPINJLMU-UHFFFAOYSA-N
di-n-octyltin oxide, stannane, dioctyloxo, dioctyltin oxide, dioctyloxostannane, tin, dioctyloxo, dioctyl oxo stannane, stannane, oxodioctyl, di-n-octyl-zinn oxyd, tin, dioctyl-, oxide, dioctyl oxo tin
93563
dioctyl(oxo)tin
CCCCCCCC[Sn](=O)CCCCCCCC

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 245°C to 248°C (decomposition)
Color White
Quantity 25 g
Formula Weight 361.14
Physical Form Powder
Chemical Name or Material Di-n-octyltin oxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'