missing translation for 'onlineSavingsMsg'
Learn More

Di-Potassium Hydrogen Orthophosphate Trihydrate, Extra Pure, SLR, Fisher Chemical™

Brand:  Fisher Chemical P/5248/60

Code : 21

Additional Details : CAS Number : 16788-57-1 Weight : 1.00000kg

Quantity 1kg

 View more versions of this product

Product Code. 10020310

Quantity Price
1 81.37 € / 1kg
4 77.28 € / 1kg
16 73.42 € / 1kg
Estimated Shipment
13-05-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Chemical Identifiers

16788-57-1
228.219
XQGPKZUNMMFTAL-UHFFFAOYSA-L
16217523
O.O.O.OP(=O)([O-])[O-].[K+].[K+]
H7K2O7P
149926
dipotassium hydrogen phosphate trihydrate, di-potassium hydrogen phosphate trihydrate, unii-rjk174x3tz, potassium hydrogenphosphate trihydrate, potassium phosphate dibasic trihydrate, potassium hydrogen phosphate trihydrate, potassium phosphate, dibasic, trihydrate, phosphoric acid, dipotassium salt, trihydrate, rjk174x3tz, dipotassium phosphate trihydrate
dipotassium;hydrogen phosphate;trihydrate

Specifications

16788-57-1
149926
XQGPKZUNMMFTAL-UHFFFAOYSA-L
dipotassium;hydrogen phosphate;trihydrate
16217523
Solid
White
1kg
H7K2O7P
dipotassium hydrogen phosphate trihydrate, di-potassium hydrogen phosphate trihydrate, unii-rjk174x3tz, potassium hydrogenphosphate trihydrate, potassium phosphate dibasic trihydrate, potassium hydrogen phosphate trihydrate, potassium phosphate, dibasic, trihydrate, phosphoric acid, dipotassium salt, trihydrate, rjk174x3tz, dipotassium phosphate trihydrate
O.O.O.OP(=O)([O-])[O-].[K+].[K+]
228.219
228.23g/mol
Plastic powder jar
8.5 to 9.6
Videos
SDS Documents