missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-Sodium Hydrogen Orthophosphate Dihydrate, Certified AR for Analysis, Fisher Chemical™
43.65 EUR valid until 2024-12-31
Use promo code "22914" to get your promotional price.
50.54 EUR valid until 2024-03-30
Use promo code "21741" to get your promotional price.
Use promo code "22914" to get your promotional price.
50.54 EUR valid until 2024-03-30
Use promo code "21741" to get your promotional price.
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Description
Di-Sodium Hydrogen Orthophosphate Dihydrate, Certified AR for Analysis, Fisher Chemical, Quantity: 500g, Packaging: Plastic powder jar, CAS: 10028-24-7, White, Melting Point: 243 deg.C, Molecular Formula: H5Na2O6P, Molecular Weight: 177.987, pH: 8.7 to 9.4
Specifications
Melting Point | 243°C |
Color | White |
pH | 8.7 to 9.4 |
Physical Form | Solid |
Quantity | 500 g |
Formula Weight | 177.99g/mol |
Packaging | Plastic powder jar |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'