Learn More
Diaion™ WK100(H), ion exchange resin, weakly acidic porous type, 2.9 meq/ml on polymethacrylate, high reaction rate, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 046647.A1
Code : NEW
Additional Details : Weight : 1.00000kg
Description
Pharmaceuticals, dealkalination, metal recovery, antibiotics, sucrose purifications
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
300-1180μm Beads | |
Weakly acidic porous type, 2.9meq/mL on polymethacrylate, high reaction rate | |
White |
Diaion™ WK100 (H), ion exchange resin | |
300 to 1180μm | |
1 kg |
RUO – Research Use Only