missing translation for 'onlineSavingsMsg'
Learn More

Dibenzo-18-crown-6, 98+%, Thermo Scientific Chemicals

Catalog Number 10357300
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
14187-32-7
C20H24O6
360.4
MFCD00005098
YSSSPARMOAYJTE-UHFFFAOYSA-N
dibenzo-18-crown-6, dibenzo-18-crown-6-ether, crown 18, 6,7,9,10,17,18,20,21-octahydrodibenzo b,k 1,4,7,10,13,16 hexaoxacyclooctadecine, dibenzo-18-crown, unii-0a7w45jcs9, 2,3:11,12-dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, dibenzo-18-crown-6 ether, dibenzo-18-crown 6-ether, 2,3,11,12-dibenzo-1,4,7,10,13,16-hexaoxacyclooctadeca-2,11-diene
26541
CHEBI:358732
C1COC2=CC=CC=C2OCCOCCOC3=CC=CC=C3OCCO1
This item is not returnable. View return policy
14187-32-7
C20H24O6
360.4
MFCD00005098
YSSSPARMOAYJTE-UHFFFAOYSA-N
dibenzo-18-crown-6, dibenzo-18-crown-6-ether, crown 18, 6,7,9,10,17,18,20,21-octahydrodibenzo b,k 1,4,7,10,13,16 hexaoxacyclooctadecine, dibenzo-18-crown, unii-0a7w45jcs9, 2,3:11,12-dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, dibenzo-18-crown-6 ether, dibenzo-18-crown 6-ether, 2,3,11,12-dibenzo-1,4,7,10,13,16-hexaoxacyclooctadeca-2,11-diene
26541
CHEBI:358732
C1COC2=CC=CC=C2OCCOCCOC3=CC=CC=C3OCCO1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY
Melting Point 160.0°C to 164.0°C
Color White to Beige
Boiling Point 380.0°C to 384.0°C (679.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Quantity 10 g
Fieser 06,159; 08,128; 09,126; 12,147
Merck Index 14, 2602
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform,toluene,ether,etc
Formula Weight 360.4
Percent Purity 98+%
Physical Form Fluffy Powder
Chemical Name or Material Dibenzo-18-crown-6, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Dibenzo-18-crown-6
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'