missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloromethane, 99.6%, ACS reagent, stabilized with amylene, Thermo Scientific Chemicals
32.03 EUR valid until 2024-03-31
Use promo code "22193" to get your promotional price.
Use promo code "22193" to get your promotional price.
Click to view available options
Quantity:
500 mL
1 L
2.5 L
25 L
Packaging:
EcoSafPak™ Glass Bottle
Glass Bottle
Metal Drum
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Melting Point | -97.0°C |
Assay Percent Range | 99.6% |
Linear Formula | CH2Cl2 |
Beilstein | 01, 60 |
Fieser | 01,676; 02,273; 04,337; 07,239; 13,106 |
Merck Index | 15, 6135 |
Solubility Information | Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol, ether and dmf |
Formula Weight | 84.93 |
Percent Purity | 99.6% |
Grade | ACS Reagent |
Mostrar más |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'